accessibility menu, dialog, popup

UserName

BrBrHNBrBr

CAS RN 55119-09-0

IUPAC Name: 1,3,6,8-tetrabromo-9H-carbazole
Synonyms: 1,3,6,8-tetrabromo-9H-carbazole
Molecular Weight (g/mol): 482.8
Molecular Formula: C12H5Br4N
InChi Key: FNHISDQCWYSMTO-UHFFFAOYSA-N
SMILES: BrC1=CC2=C(NC3=C2C=C(Br)C=C3Br)C(Br)=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results