accessibility menu, dialog, popup

UserName

CAS RN 5500-21-0

IUPAC Name: cyclopropanecarbonitrile
Synonyms: cyclopropyl cyanide
Molecular Weight (g/mol): 67.09
Molecular Formula: C4H5N
InChi Key: AUQDITHEDVOTCU-UHFFFAOYSA-N
SMILES: N#CC1CC1
Boiling Point 
  • (1)
  • (3)
  • (2)
Molecular Weight (g/mol) 
  • (1)
  • (5)
Percent Purity 
  • (2)
  • (4)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (4)
  • (1)
  • (3)
  • (2)
  • (1)
  • (5)
  • (2)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)