accessibility menu, dialog, popup

UserName

CH3SNHNOOH3CHN

CAS RN 54965-21-8

IUPAC Name: methyl N-[6-(propylsulfanyl)-1H-1,3-benzodiazol-2-yl]carbamate
Synonyms: albendazol disthelm, mediamix V disthelm, mediamix v sanicoopa brand of albendazole mediamix v disthelm smith kline & french brand of albendazole disthelm albendazole pfizer brand bilutac bendapar
Molecular Weight (g/mol): 265.33
Molecular Formula: C12H15N3O2S
InChi Key: HXHWSAZORRCQMX-UHFFFAOYSA-N
SMILES: CCCSC1=CC=C2N=C(NC(=O)OC)NC2=C1
Percent Purity 
  • (4)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Grade 
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (2)
  • (3)
  • (4)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)