missing translation for 'onlineSavingsMsg'
Learn More

Albendazole 98.0+%, TCI America™

Catalog No. A19435G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A19435G 5 g
A194325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A19435G Supplier TCI America Supplier No. A19435G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 54965-21-8
Molecular Formula C12H15N3O2S
Molecular Weight (g/mol) 265.33
MDL Number MFCD00083232
InChI Key HXHWSAZORRCQMX-UHFFFAOYSA-N
Synonym albendazole, albenza, eskazole, valbazen, zentel, albendazol, proftril, albendazolum, bilutac, zental
PubChem CID 2082
ChEBI CHEBI:16664
IUPAC Name methyl N-[6-(propylsulfanyl)-1H-1,3-benzodiazol-2-yl]carbamate
SMILES CCCSC1=CC=C2N=C(NC(=O)OC)NC2=C1

Specifications

Melting Point 210°C
Color White-Yellow
Quantity 5 g
Formula Weight 265.33
Percent Purity ≥98.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material Albendazole
RTECSNumber FD1100000
TSCA No