missing translation for 'onlineSavingsMsg'
Learn More

Albendazole, 98%, MilliporeSigma™ Supelco™

Catalog No. 111014582
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111014582 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111014582 Supplier Merck Emd Millipore Supplier No. A467310G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Albendazole is a benzimidazole derivative used for the treatment of human gastrointestinal helmintiasis. It inhibits microtubule and blocks the glucose uptake that causes depletion of glycogen stores and lowering formation of adinosine triphosphate (ATP) in the larval and adult stages of susceptible parasite. Featured Industries: Pharmaceutical (small molecule)

Specifications

CAS 54965-21-8
Physical Form Neat
Linear Formula C12H15N3O2 S
MDL Number MFCD00083232
Synonym Methyl 5-(propylthio)-2-benzimidazolecarbamate
Type Analytical Standard
Quantity 10 g
Formula Weight 265.33
Percent Purity ≥98%
Grade Analytical Standard