accessibility menu, dialog, popup

UserName

CAS RN 54593-26-9

IUPAC Name: 3,5-dimethyl-1,2-oxazole-4-carbaldehyde
Synonyms: 3,5-dimethyl-1,2-oxazole-4-carbaldehyde
Molecular Weight (g/mol): 125.13
Molecular Formula: C6H7NO2
InChi Key: TVAYXKLCEILMEA-UHFFFAOYSA-N
SMILES: CC1=C(C=O)C(C)=NO1
Percent Purity 
  • (1)
  • (5)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (5)
  • (1)
  • (5)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)