missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dimethylisoxazole-4-carboxaldehyde 98.0+%, TCI America™

Catalog No. D46821G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D46821G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D46821G Supplier TCI America Supplier No. D46821G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 54593-26-9
Molecular Formula C6H7NO2
Molecular Weight (g/mol) 125.127
MDL Number MFCD02681977
InChI Key TVAYXKLCEILMEA-UHFFFAOYSA-N
Synonym 3,5-dimethyl-4-isoxazolecarbaldehyde, 3,5-dimethylisoxazole-4-carbaldehyde, 3,5-dimethylisoxazole-4-carboxaldehyde, 4-isoxazolecarboxaldehyde, 3,5-dimethyl, 3,5-dimethyl-4-isoxazolecarboxaldehyde, 3,5-dimethyl-4-formalisoxazole, pubchem8682, acmc-1axd1, ksc496e3j, 3,5-dimethylisoxazol-4-carbaldehyde
PubChem CID 289576
IUPAC Name 3,5-dimethyl-1,2-oxazole-4-carbaldehyde
SMILES CC1=C(C(=NO1)C)C=O

Specifications

Color White
Boiling Point 73°C
Quantity 1 g
Formula Weight 125.13
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3,5-Dimethylisoxazole-4-carboxaldehyde