accessibility menu, dialog, popup

UserName

ClNClHNNN

CAS RN 5451-40-1

IUPAC Name: 2,6-dichloro-7H-purine
Synonyms: 2,6-dichloro-7H-purine
Molecular Weight (g/mol): 189
Molecular Formula: C5H2Cl2N4
InChi Key: RMFWVOLULURGJI-UHFFFAOYSA-N
SMILES: ClC1=NC(Cl)=C2NC=NC2=N1
Percent Purity 
  • (3)
  • (4)
Quantity 
  • (3)
  • (1)
  • (3)
Physical Form 
  • (5)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (3)
  • (4)
  • (3)
  • (4)
  • (3)
  • (1)
  • (3)
  • (5)