missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dichloropurine, 97%

Catalog No. AAB2128906
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2128906 5 g
AAB2128903 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2128906 Supplier Thermo Scientific Chemicals Supplier No. B2128906
Get Up to $1,800 in Chemicals with Purchase
View all promotions

missing translation for 'validMainframeMsg'

CAS: 5451-40-1 | C5H2Cl2N4 | 189.00 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 5451-40-1
Molecular Formula C5H2Cl2N4
Molecular Weight (g/mol) 189.00
MDL Number MFCD00077725
InChI Key RMFWVOLULURGJI-UHFFFAOYSA-N
Synonym 2,6-dichloropurine, 2,6-dichloro-9h-purine, 2,6-dichloro-1h-purine, 9h-purine, 2,6-dichloro, 2,6-dichloro purine, 2,6 dichloropurine, 2,6-dichioropurine, purine,6-dichloro, zlchem 226, 2,6-dichloro-purine
PubChem CID 5324412
IUPAC Name 2,6-dichloro-7H-purine
SMILES ClC1=NC(Cl)=C2NC=NC2=N1

Specifications

Melting Point ∼184°C to 186°C (decomposition)
Quantity 5 g
Beilstein 9354
Formula Weight 189.01
Percent Purity 97%
Chemical Name or Material 2,6-Dichloropurine
Hazard Category H301-H315-H319-H335
Precautionary Statement P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 226-681-6
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only