accessibility menu, dialog, popup

UserName

CH3OHN

CAS RN 5436-83-9

IUPAC Name: N-benzyl-4-methylbenzamide
Synonyms: N-benzyl-4-methylbenzamide
Molecular Weight (g/mol): 225.29
Molecular Formula: C15H15NO
InChi Key: QIULRTIURUDCGH-UHFFFAOYSA-N
SMILES: CC1=CC=C(C=C1)C(=O)NCC1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results