missing translation for 'onlineSavingsMsg'
Learn More

N-Benzyl-4-methylbenzamide, 97%, Thermo Scientific™

Catalog No. AAH56270MD
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH56270MD 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH56270MD Supplier Thermo Scientific Chemicals Supplier No. H56270MD
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff. 

Notes
Store in a cool dry place. Incompatible with oxidizing agents. Keep container tightly closed.

Chemical Identifiers

CAS 5436-83-9
Molecular Formula C15H15NO
Molecular Weight (g/mol) 225.291
MDL Number MFCD00026008
InChI Key QIULRTIURUDCGH-UHFFFAOYSA-N
Synonym benzamide, 4-methyl-n-phenylmethyl, cambridge id 5104261, benzamide,4-methyl-n-phenylmethyl
PubChem CID 228681
IUPAC Name N-benzyl-4-methylbenzamide
SMILES CC1=CC=C(C=C1)C(=O)NCC2=CC=CC=C2

Specifications

Quantity 250 mg
Formula Weight 225.29
Percent Purity 97%
Chemical Name or Material N-Benzyl-4-methylbenzamide
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures