CAS RN 5427-26-9
IUPAC Name:
2-(2,5-dioxoimidazolidin-4-yl)acetic acid
Synonyms:
(2,5-dioxoimidazolidin-4-yl)acetic acid
Molecular Weight (g/mol):
158.11
Molecular Formula:
C5H6N2O4
InChi Key:
DQQLZADYSWBCOX-UHFFFAOYNA-N
SMILES:
OC(=O)CC1NC(=O)NC1=O
1
–
1
of
1
results
1
Promotions Available
Hydantoin-5-acetic acid, 98%
Hydantoin-5-acetic acid, 98%
CAS: 5427-26-9 Molecular Formula: C5H6N2O4 Molecular Weight (g/mol): 158.11 MDL Number: MFCD00005267 InChI Key: DQQLZADYSWBCOX-UHFFFAOYNA-N Synonym: hydantoin-5-acetic acid,5-hydantoinacetic acid,2,5-dioxoimidazolidin-4-yl acetic acid,5-hydantoin acetic acid,2-2,5-dioxoimidazolidin-4-yl acetic acid,4-imidazolidineacetic acid, 2,5-dioxo,2,5-dioxo-1,3-diazolidin-4-ylacetic acid,2-2,5-dioxo-1,3-diazolidin-4-yl acetic acid,5-carboxymethylhydantoin,acmc-20akr6 PubChem CID: 95492 IUPAC Name: 2-(2,5-dioxoimidazolidin-4-yl)acetic acid SMILES: OC(=O)CC1NC(=O)NC1=O