missing translation for 'onlineSavingsMsg'
Learn More

Hydantoin-5-acetic acid, 98%

Catalog No. AAA1173114
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1173114 25 g
AAA1173106 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1173114 Supplier Thermo Scientific Chemicals Supplier No. A1173114
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 5427-26-9 | C5H6N2O4 | 158.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 5427-26-9
Molecular Formula C5H6N2O4
Molecular Weight (g/mol) 158.11
MDL Number MFCD00005267
InChI Key DQQLZADYSWBCOX-UHFFFAOYNA-N
Synonym hydantoin-5-acetic acid, 5-hydantoinacetic acid, 2,5-dioxoimidazolidin-4-yl acetic acid, 5-hydantoin acetic acid, 2-2,5-dioxoimidazolidin-4-yl acetic acid, 4-imidazolidineacetic acid, 2,5-dioxo, 2,5-dioxo-1,3-diazolidin-4-ylacetic acid, 2-2,5-dioxo-1,3-diazolidin-4-yl acetic acid, 5-carboxymethylhydantoin, acmc-20akr6
PubChem CID 95492
IUPAC Name 2-(2,5-dioxoimidazolidin-4-yl)acetic acid
SMILES OC(=O)CC1NC(=O)NC1=O

Specifications

Melting Point ∼217°C (decomposition)
Quantity 25 g
Beilstein 83035
Formula Weight 158.11
Percent Purity 98%
Chemical Name or Material Hydantoin-5-acetic acid
Hazard Category Warning
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
EINECSNumber 226-574-4
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only