accessibility menu, dialog, popup

UserName

(S)(R)(S)(R)O(R)(R)O(S)(S)(S)(S)(S)OOO(S)(S)(R)(R)(R)(R)(R)(R)OOHOOHOHOHHOHOHOOHHOHOHOHOOHHO

CAS RN 54261-98-2

IUPAC Name: (2R,3R,4S,5S,6R)-2-{[(2S,3S,4S,5R)-3,4-dihydroxy-2,5-bis(hydroxymethyl)oxolan-2-yl]oxy}-6-({[(2S,3R,4S,5R,6R)-3,4,5-trihydroxy-6-({[(2S,3R,4S,5R,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}methyl)oxan-2-yl]oxy}methyl)oxane-3,4,5-triol
Synonyms: stachyose
Molecular Weight (g/mol): 666.58
Molecular Formula: C24H42O21
InChi Key: UQZIYBXSHAGNOE-CYDMKDQUNA-N
SMILES: OC[C@H]1O[C@@](CO)(O[C@H]2O[C@H](CO[C@H]3O[C@H](CO[C@H]4O[C@H](CO)[C@H](O)[C@H](O)[C@H]4O)[C@H](O)[C@H](O)[C@H]3O)[C@@H](O)[C@H](O)[C@H]2O)[C@@H](O)[C@@H]1O
Quantity 
  • (3)
  • (1)
  • (3)
  • (1)
Percent Purity 
  • (3)
  • (2)
Grade 
  • (3)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results