missing translation for 'onlineSavingsMsg'
Learn More

Stachyose hydrate, technical

Catalog No. AC453142500
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC453142500 250 mg
AC453140010 1 g
AC453140050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC453142500 Supplier Thermo Scientific Chemicals Supplier No. 453142500
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 54261-98-2 | C24H42O21 | 666.58 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 54261-98-2
Molecular Formula C24H42O21
Molecular Weight (g/mol) 666.58
MDL Number MFCD00149457
InChI Key UQZIYBXSHAGNOE-CYDMKDQUNA-N
PubChem CID 131801001
SMILES OC[C@H]1O[C@@](CO)(O[C@H]2O[C@H](CO[C@H]3O[C@H](CO[C@H]4O[C@H](CO)[C@H](O)[C@H](O)[C@H]4O)[C@H](O)[C@H](O)[C@H]3O)[C@@H](O)[C@H](O)[C@H]2O)[C@@H](O)[C@@H]1O

Specifications

Melting Point 95.0°C to 105.0°C
Infrared Spectrum Authentic
Quantity 250 mg
Specific Rotation Condition + 129.00 (20.00°C c=4, H2O)
Specific Rotation + 129.00
Solubility Information Solubility in water: soluble.
IUPAC Name (2R,3R,4S,5S,6R)-2-{[(2S,3S,4S,5R)-3,4-dihydroxy-2,5-bis(hydroxymethyl)oxolan-2-yl]oxy}-6-({[(2S,3R,4S,5R,6R)-3,4,5-trihydroxy-6-({[(2S,3R,4S,5R,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}methyl)oxan-2-yl]oxy}methyl)oxane-3,4,5-triol
Formula Weight 666.58
Grade Technical
Chemical Name or Material Stachyose hydrate

RUO – Research Use Only