accessibility menu, dialog, popup

UserName

CAS RN 5426-55-1

NaH3CNH3COHO

CAS RN 5426-55-1

IUPAC Name: 2-(diethylamino)acetic acid sodium
Synonyms: N,N-diethylglycine sodium
Molecular Weight (g/mol): 154.17
Molecular Formula: C6H13NNaO2
InChi Key: XZQDOIKSUNQDGV-UHFFFAOYSA-N
SMILES: [Na].CCN(CC)CC(O)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results