accessibility menu, dialog, popup

UserName

CAS RN 542-85-8

IUPAC Name: isothiocyanatoethane
Synonyms: isothiocyanatoethane
Molecular Weight (g/mol): 87.14
Molecular Formula: C3H5NS
InChi Key: HBNYJWAFDZLWRS-UHFFFAOYSA-N
SMILES: CCN=C=S
Boiling Point 
  • (6)
  • (1)
Percent Purity 
  • (1)
  • (3)
  • (3)
Quantity 
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (1)
  • (6)
  • (6)
  • (1)
  • (1)
  • (3)
  • (3)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (4)