missing translation for 'onlineSavingsMsg'
Learn More

Ethyl Isothiocyanate 97.0+%, TCI America™

Catalog No. I018825ML
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I018825ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. I018825ML Supplier TCI America Supplier No. I018825ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 542-85-8
Molecular Formula C3H5NS
Molecular Weight (g/mol) 87.14
MDL Number MFCD00004820
InChI Key HBNYJWAFDZLWRS-UHFFFAOYSA-N
Synonym ethyl isothiocyanate, ethane, isothiocyanato, ethyl mustard oil, ethylisothiocyanate, isothiocyanic acid, ethyl ester, unii-3284mj2t8p, ccris 7323, ethylisothio-cyanate, isothiocyanato-ethane
PubChem CID 10966
ChEBI CHEBI:85098
IUPAC Name isothiocyanatoethane
SMILES CCN=C=S

Specifications

Color Yellow
Boiling Point 132°C
Quantity 25 mL
UN Number 1992
Formula Weight 87.14
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethyl Isothiocyanate
RTECSNumber NX8590000
TSCA Yes