accessibility menu, dialog, popup

UserName

CAS RN 542-10-9

CH3OH3COOCH3O

CAS RN 542-10-9

IUPAC Name: 1-(acetyloxy)ethyl acetate
Synonyms: ethylidene diacetate
Molecular Weight (g/mol): 146.14
Molecular Formula: C6H10O4
InChi Key: ACKALUBLCWJVNB-UHFFFAOYSA-N
SMILES: CC(OC(C)=O)OC(C)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results