missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,1-Ethanediol Diacetate 98.0+%, TCI America™
Specifications
Specifications
| Color | Yellow |
| Boiling Point | 167°C |
| Quantity | 25 g |
| Formula Weight | 146.14 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Liquid |
| Chemical Name or Material | 1,1-Ethanediol Diacetate |
Safety and Handling
| TSCA | Yes |