accessibility menu, dialog, popup

UserName

H3CBrCH3Br

CAS RN 5408-86-6

IUPAC Name: 2,3-dibromobutane
Synonyms: 2,3-dibromobutane
Molecular Weight (g/mol): 215.92
Molecular Formula: C4H8Br2
InChi Key: BXXWFOGWXLJPPA-UHFFFAOYNA-N
SMILES: CC(Br)C(C)Br
Boiling Point 
  • (1)
  • (3)
Percent Purity 
  • (3)
  • (1)
Quantity 
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (3)
  • (1)
  • (3)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)