accessibility menu, dialog, popup

UserName

ONOHNN

CAS RN 5401-94-5

IUPAC Name: 5-nitro-1H-indazole
Synonyms: 1H-indazole, 5-nitro-
Molecular Weight (g/mol): 163.14
Molecular Formula: C7H5N3O2
InChi Key: WSGURAYTCUVDQL-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC=C2NN=CC2=C1
Percent Purity 
  • (3)
  • (1)
Quantity 
  • (1)
  • (2)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (3)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)