missing translation for 'onlineSavingsMsg'
Learn More

5-Nitroindazole 98.0+%, TCI America™

Catalog No. N039825G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N039825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N039825G Supplier TCI America Supplier No. N039825G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 5401-94-5
Molecular Formula C7H5N3O2
Molecular Weight (g/mol) 163.136
MDL Number MFCD00005693
InChI Key WSGURAYTCUVDQL-UHFFFAOYSA-N
Synonym 5-nitroindazole, 1h-indazole, 5-nitro, unii-235y7p37zd, ccris 4134, 2h-indazole, 5-nitro, 5ni, 5-nitroindazol, 5-nitro indazole, 5-nitro-indazole, 5-nitro-1h-indazol
PubChem CID 21501
IUPAC Name 5-nitro-1H-indazole
SMILES C1=CC2=C(C=C1[N+](=O)[O-])C=NN2

Specifications

Melting Point 210°C
Quantity 25 g
Formula Weight 163.14
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 5-Nitroindazole
EINECSNumber (5)-0270
RTECSNumber NK7962000
TSCA Yes