CAS RN 540-84-1
Isooctane, Optima™ for HPLC and GC, Fisher Chemical™
CAS: 540-84-1 Molecular Formula: C8H18 Molecular Weight (g/mol): 114.232 InChI Key: NHTMVDHEPJAVLT-UHFFFAOYSA-N Synonym: isooctane,isobutyltrimethylmethane,pentane, 2,2,4-trimethyl,iso-octane,2,4,4-trimethylpentane,unii-qab8f5669o,ch3 2chch2c ch3 3,2, 2, 4-trimethylpentane,i-octane,isobutyltrimethylethane PubChem CID: 10907 ChEBI: CHEBI:62805 IUPAC Name: 2,2,4-trimethylpentane SMILES: CC(C)CC(C)(C)C
Isooctane (HPLC), Fisher Chemical™
CAS: 540-84-1 Molecular Formula: C8H18 Molecular Weight (g/mol): 114.232 MDL Number: MFCD00008943 InChI Key: NHTMVDHEPJAVLT-UHFFFAOYSA-N Synonym: isooctane,isobutyltrimethylmethane,pentane, 2,2,4-trimethyl,iso-octane,2,4,4-trimethylpentane,unii-qab8f5669o,ch3 2chch2c ch3 3,2, 2, 4-trimethylpentane,i-octane,isobutyltrimethylethane PubChem CID: 10907 ChEBI: CHEBI:62805 IUPAC Name: 2,2,4-trimethylpentane SMILES: CC(C)CC(C)(C)C
Isooctane (Certified ACS), Fisher Chemical™
CAS: 540-84-1 Molecular Formula: C8H18 Molecular Weight (g/mol): 114.232 MDL Number: MFCD00008943 InChI Key: NHTMVDHEPJAVLT-UHFFFAOYSA-N Synonym: isooctane,isobutyltrimethylmethane,pentane, 2,2,4-trimethyl,iso-octane,2,4,4-trimethylpentane,unii-qab8f5669o,ch3 2chch2c ch3 3,2, 2, 4-trimethylpentane,i-octane,isobutyltrimethylethane PubChem CID: 10907 ChEBI: CHEBI:62805 IUPAC Name: 2,2,4-trimethylpentane SMILES: CC(C)CC(C)(C)C
Isooctane (Spectranalyzed™), Fisher Chemical™
CAS: 540-84-1 Molecular Formula: C8H18 Molecular Weight (g/mol): 114.232 MDL Number: MFCD00008943 InChI Key: NHTMVDHEPJAVLT-UHFFFAOYSA-N Synonym: isooctane,isobutyltrimethylmethane,pentane, 2,2,4-trimethyl,iso-octane,2,4,4-trimethylpentane,unii-qab8f5669o,ch3 2chch2c ch3 3,2, 2, 4-trimethylpentane,i-octane,isobutyltrimethylethane PubChem CID: 10907 ChEBI: CHEBI:62805 IUPAC Name: 2,2,4-trimethylpentane SMILES: CC(C)CC(C)(C)C
Isooctane (Pesticide), Fisher Chemical
CAS: 540-84-1 Molecular Formula: C8H18 Molecular Weight (g/mol): 114.232 MDL Number: MFCD00008943 InChI Key: NHTMVDHEPJAVLT-UHFFFAOYSA-N Synonym: isooctane,isobutyltrimethylmethane,pentane, 2,2,4-trimethyl,iso-octane,2,4,4-trimethylpentane,unii-qab8f5669o,ch3 2chch2c ch3 3,2, 2, 4-trimethylpentane,i-octane,isobutyltrimethylethane PubChem CID: 10907 ChEBI: CHEBI:62805 IUPAC Name: 2,2,4-trimethylpentane SMILES: CC(C)CC(C)(C)C
2,2,4-Trimethylpentane, 99+%, pure
CAS: 540-84-1 Molecular Formula: C8H18 Molecular Weight (g/mol): 114.232 MDL Number: MFCD00008943 InChI Key: NHTMVDHEPJAVLT-UHFFFAOYSA-N Synonym: isooctane,isobutyltrimethylmethane,pentane, 2,2,4-trimethyl,iso-octane,2,4,4-trimethylpentane,unii-qab8f5669o,ch3 2chch2c ch3 3,2, 2, 4-trimethylpentane,i-octane,isobutyltrimethylethane PubChem CID: 10907 ChEBI: CHEBI:62805 IUPAC Name: 2,2,4-trimethylpentane SMILES: CC(C)CC(C)(C)C
2,2,4-Trimethylpentane, 99%
CAS: 540-84-1 Molecular Formula: C8H18 Molecular Weight (g/mol): 114.232 MDL Number: MFCD00008943 InChI Key: NHTMVDHEPJAVLT-UHFFFAOYSA-N Synonym: isooctane,isobutyltrimethylmethane,pentane, 2,2,4-trimethyl,iso-octane,2,4,4-trimethylpentane,unii-qab8f5669o,ch3 2chch2c ch3 3,2, 2, 4-trimethylpentane,i-octane,isobutyltrimethylethane PubChem CID: 10907 ChEBI: CHEBI:62805 IUPAC Name: 2,2,4-trimethylpentane SMILES: CC(C)CC(C)(C)C
2,2,4-Trimethylpentane, HPLC Grade, 99.7+%
CAS: 540-84-1 Molecular Formula: C8H18 Molecular Weight (g/mol): 114.232 MDL Number: MFCD00008943 InChI Key: NHTMVDHEPJAVLT-UHFFFAOYSA-N Synonym: isooctane,isobutyltrimethylmethane,pentane, 2,2,4-trimethyl,iso-octane,2,4,4-trimethylpentane,unii-qab8f5669o,ch3 2chch2c ch3 3,2, 2, 4-trimethylpentane,i-octane,isobutyltrimethylethane PubChem CID: 10907 ChEBI: CHEBI:62805 IUPAC Name: 2,2,4-trimethylpentane SMILES: CC(C)CC(C)(C)C
2,2,4-Trimethylpentane, ACS, 99+%
CAS: 540-84-1 Molecular Formula: C8H18 Molecular Weight (g/mol): 114.232 MDL Number: MFCD00008943 InChI Key: NHTMVDHEPJAVLT-UHFFFAOYSA-N Synonym: isooctane,isobutyltrimethylmethane,pentane, 2,2,4-trimethyl,iso-octane,2,4,4-trimethylpentane,unii-qab8f5669o,ch3 2chch2c ch3 3,2, 2, 4-trimethylpentane,i-octane,isobutyltrimethylethane PubChem CID: 10907 ChEBI: CHEBI:62805 IUPAC Name: 2,2,4-trimethylpentane SMILES: CC(C)CC(C)(C)C
Lead in Isooctane standard solution, Specpure™, 37μg/g(0.1g/gal), Thermo Scientific Chemicals
CAS: 540-84-1 Molecular Formula: C8H18 Molecular Weight (g/mol): 114.232 MDL Number: MFCD01094200 InChI Key: NHTMVDHEPJAVLT-UHFFFAOYSA-N PubChem CID: 10907 ChEBI: CHEBI:62805 IUPAC Name: 2,2,4-trimethylpentane SMILES: CC(C)CC(C)(C)C
2,2,4-Trimethylpentane, 99.5%, for residue analysis, ECD tested for pesticide analysis
CAS: 540-84-1 Molecular Formula: C8H18 Molecular Weight (g/mol): 114.232 MDL Number: MFCD00008943 InChI Key: NHTMVDHEPJAVLT-UHFFFAOYSA-N Synonym: isooctane,isobutyltrimethylmethane,pentane, 2,2,4-trimethyl,iso-octane,2,4,4-trimethylpentane,unii-qab8f5669o,ch3 2chch2c ch3 3,2, 2, 4-trimethylpentane,i-octane,isobutyltrimethylethane PubChem CID: 10907 ChEBI: CHEBI:62805 IUPAC Name: 2,2,4-trimethylpentane SMILES: CC(C)CC(C)(C)C
Iso-Octane, ACS Reagent, for organic synthesis, prep-LC, and general laboratory use, >99.0%, Solstice
CAS: 540-84-1 Molecular Formula: C8H18 Molecular Weight (g/mol): 114.232 InChI Key: NHTMVDHEPJAVLT-UHFFFAOYSA-N Synonym: isooctane,isobutyltrimethylmethane,pentane, 2,2,4-trimethyl,iso-octane,2,4,4-trimethylpentane,unii-qab8f5669o,ch3 2chch2c ch3 3,2, 2, 4-trimethylpentane,i-octane,isobutyltrimethylethane PubChem CID: 10907 ChEBI: CHEBI:62805 IUPAC Name: 2,2,4-trimethylpentane SMILES: CC(C)CC(C)(C)C
2,2,4-Trimethylpentane, 99+%, ACS reagent
CAS: 540-84-1 Molecular Formula: C8H18 Molecular Weight (g/mol): 114.232 MDL Number: MFCD00008943 InChI Key: NHTMVDHEPJAVLT-UHFFFAOYSA-N Synonym: isooctane,isobutyltrimethylmethane,pentane, 2,2,4-trimethyl,iso-octane,2,4,4-trimethylpentane,unii-qab8f5669o,ch3 2chch2c ch3 3,2, 2, 4-trimethylpentane,i-octane,isobutyltrimethylethane PubChem CID: 10907 ChEBI: CHEBI:62805 IUPAC Name: 2,2,4-trimethylpentane SMILES: CC(C)CC(C)(C)C
2,2,4-Trimethylpentane, Pesticide Residueanalysis Grade, Thermo Scientific Chemicals
CAS: 540-84-1 Molecular Formula: C8H18 Molecular Weight (g/mol): 114.232 MDL Number: MFCD00008943 InChI Key: NHTMVDHEPJAVLT-UHFFFAOYSA-N Synonym: isooctane,isobutyltrimethylmethane,pentane, 2,2,4-trimethyl,iso-octane,2,4,4-trimethylpentane,unii-qab8f5669o,ch3 2chch2c ch3 3,2, 2, 4-trimethylpentane,i-octane,isobutyltrimethylethane PubChem CID: 10907 ChEBI: CHEBI:62805 IUPAC Name: 2,2,4-trimethylpentane SMILES: CC(C)CC(C)(C)C