accessibility menu, dialog, popup

UserName

CAS RN 540-38-5

IUPAC Name: 4-iodophenol
Synonyms: 4-iodophenol 4-jodphenol p-jodphenol ccris 668
Molecular Weight (g/mol): 220.01
Molecular Formula: C6H5IO
InChi Key: VSMDINRNYYEDRN-UHFFFAOYSA-N
SMILES: OC1=CC=C(I)C=C1
Percent Purity 
  • (4)
  • (4)
  • (3)
Quantity 
  • (1)
  • (2)
  • (2)
  • (3)
  • (2)
  • (1)
Boiling Point 
  • (4)
  • (3)
Physical Form 
  • (4)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (7)
  • (4)
  • (7)
  • (4)
  • (4)
  • (3)
  • (1)
  • (2)
  • (2)
  • (3)
  • (2)
  • (1)
  • (4)
  • (3)
  • (4)
  • (3)