accessibility menu, dialog, popup

UserName

OHHNOO

CAS RN 54-16-0

IUPAC Name: 2-(5-hydroxy-1H-indol-3-yl)acetate
Synonyms: 5-hydroxyindoleacetate 5-hydroxyindoleacetate(1-)
Molecular Weight (g/mol): 190.18
Molecular Formula: C10H8NO3
InChi Key: DUUGKQCEGZLZNO-UHFFFAOYSA-M
SMILES: OC1=CC=C2NC=C(CC([O-])=O)C2=C1
Percent Purity 
  • (1)
  • (2)
  • (2)
  • (3)
Quantity 
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Molecular Weight (g/mol) 
  • (4)
  • (3)
Physical Form 
  • (7)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (2)
  • (5)
  • (1)
  • (2)
  • (2)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (4)
  • (3)
  • (7)
  • (1)