accessibility menu, dialog, popup

UserName

CAS RN 53924-05-3

IUPAC Name: 7-chloro-1H-indole
Synonyms: indole-7-chloro
Molecular Weight (g/mol): 151.59
Molecular Formula: C8H6ClN
InChi Key: WMYQAKANKREQLM-UHFFFAOYSA-N
SMILES: ClC1=C2NC=CC2=CC=C1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)