accessibility menu, dialog, popup

UserName

CAS RN 5392-28-9

IUPAC Name: pyrimidine-2,4,5,6-tetramine; sulfuric acid
Synonyms: tetraaminopyrimidine sulfate
Molecular Weight (g/mol): 238.22
Molecular Formula: C4H10N6O4S
InChi Key: MQEFDQWUCTUJCP-UHFFFAOYSA-N
SMILES: OS(O)(=O)=O.NC1=NC(N)=C(N)C(N)=N1
Percent Purity 
  • (1)
  • (3)
  • (2)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (1)
  • (5)
  • (1)
  • (3)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (3)