missing translation for 'onlineSavingsMsg'
Learn More

2,4,5,6-Tetraaminopyrimidine Sulfate 98.0+%, TCI America™

Catalog No. T146225G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T146225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T146225G Supplier TCI America Supplier No. T146225G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 5392-28-9
Molecular Formula C4H10N6O4S
Molecular Weight (g/mol) 238.22
MDL Number MFCD06408008
InChI Key MQEFDQWUCTUJCP-UHFFFAOYSA-N
Synonym 2,4,5,6-tetraaminopyrimidine sulfate, pyrimidinetetramine sulfate, pyrimidine-2,4,5,6-tetraamine sulfate, pyrimidine-2,4,5,6-tetramine; sulfuric acid, tetraaminopyrimidine sulfate, pyrimidinetetramine, sulfate 1:1, pyrimidinetetramine, sulfate, tetraaminopyrimidine sulphate, unii-53qqy0i99m, tetra-aminopyrimidine sulfate
PubChem CID 79358
IUPAC Name pyrimidine-2,4,5,6-tetramine; sulfuric acid
SMILES OS(O)(=O)=O.NC1=NC(N)=C(N)C(N)=N1

Specifications

Melting Point 300°C
Quantity 25 g
Formula Weight 238.22
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 2,4,5,6-Tetraaminopyrimidine Sulfate
RTECSNumber UV9738000
TSCA Yes