accessibility menu, dialog, popup

UserName

H3COBrOOCH3

CAS RN 5392-10-9

IUPAC Name: 2-bromo-4,5-dimethoxybenzaldehyde
Synonyms: 2-bromo-4,5-dimethoxybenzaldehyde
Molecular Weight (g/mol): 245.07
Molecular Formula: C9H9BrO3
InChi Key: UQQROBHFUDBOOK-UHFFFAOYSA-N
SMILES: COC1=CC(Br)=C(C=O)C=C1OC
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (1)
  • (2)
  • (2)
  • (2)