CAS RN 53916-94-2
IUPAC Name:
2-phenylethan-1-amine hydrobromide
Synonyms:
phenethylamine hydrobromide
Molecular Weight (g/mol):
202.1
Molecular Formula:
C8H12BrN
InChi Key:
IRAGENYJMTVCCV-UHFFFAOYSA-N
SMILES:
Br.NCCC1=CC=CC=C1
1
–
1
of
1
results
1
2-Phenylethylamine Hydrobromide 98.0+%, TCI America™
CAS: 53916-94-2 Molecular Formula: C8H12BrN Molecular Weight (g/mol): 202.095 InChI Key: IRAGENYJMTVCCV-UHFFFAOYSA-N Synonym: Phenethylamine Hydrobromide, 2-Phenylethylammonium Bromide PubChem CID: 70441016 IUPAC Name: 2-phenylethanamine;hydrobromide SMILES: C1=CC=C(C=C1)CCN.Br