accessibility menu, dialog, popup

UserName

H3COOCH3

CAS RN 539-82-2

IUPAC Name: ethyl pentanoate
Synonyms: ethyl valerate
Molecular Weight (g/mol): 130.19
Molecular Formula: C7H14O2
InChi Key: ICMAFTSLXCXHRK-UHFFFAOYSA-N
SMILES: CCCCC(=O)OCC
Boiling Point 
  • (2)
  • (5)
Molecular Weight (g/mol) 
  • (4)
  • (3)
Percent Purity 
  • (2)
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (5)
  • (2)
  • (5)
  • (4)
  • (3)
  • (2)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (5)