missing translation for 'onlineSavingsMsg'
Learn More

Ethyl valerate, 98%

Catalog No. AAA14854AP
Change view
Click to view available options
Quantity:
500 mL
2500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA14854AP 500 mL
AAA148540F 2500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA14854AP Supplier Thermo Scientific Chemicals Supplier No. A14854AP
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 539-82-2 | C7H14O2 | 130.187 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 539-82-2
Molecular Formula C7H14O2
Molecular Weight (g/mol) 130.187
MDL Number MFCD00009479
InChI Key ICMAFTSLXCXHRK-UHFFFAOYSA-N
Synonym ethyl valerate, ethyl n-valerate, pentanoic acid, ethyl ester, valeric acid, ethyl ester, ethylvalerate, ethyl valerianate, pentanoic acid ethyl ester, n-valeric acid ethyl ester, valeric acid, ethyl ester 8ci, fema no. 2462
PubChem CID 10882
IUPAC Name ethyl pentanoate
SMILES CCCCC(=O)OCC

Specifications

Melting Point -92°C to -90°C
Density 0.874
Boiling Point 142°C to 145°C
Flash Point 38°C (100°F)
Refractive Index 1.4
Quantity 500 mL
UN Number UN3272
Beilstein 1744680
Merck Index 14,9904
Formula Weight 130.19
Percent Purity 98%
Chemical Name or Material Ethyl valerate
Show More Show Less
Hazard Category H226
Hazard Statement GHS H Statement
H226
Flammable liquid and vapour.
Precautionary Statement P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ESTERS, N.O.S.
EINECSNumber 208-726-1
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only