accessibility menu, dialog, popup

UserName

CAS RN 539-03-7

IUPAC Name: N-(4-chlorophenyl)acetamide
Synonyms: 4-chloroacetanilide P-chloroacetanilide
Molecular Weight (g/mol): 169.61
Molecular Formula: C8H8ClNO
InChi Key: GGUOCFNAWIODMF-UHFFFAOYSA-N
SMILES: CC(=O)NC1=CC=C(Cl)C=C1
Molecular Weight (g/mol) 
  • (5)
  • (3)
Percent Purity 
  • (3)
  • (2)
  • (3)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
Physical Form 
  • (5)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (2)
  • (6)
  • (5)
  • (3)
  • (3)
  • (2)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (6)
  • (5)