accessibility menu, dialog, popup

UserName

N(E/Z)

CAS RN 538-51-2

IUPAC Name: N,1-diphenylmethanimine
Synonyms: benzalaniline
Molecular Weight (g/mol): 181.24
Molecular Formula: C13H11N
InChi Key: UVEWQKMPXAHFST-UHFFFAOYSA-N
SMILES: C(=NC1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (2)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)