missing translation for 'onlineSavingsMsg'
Learn More

N-Benzylideneaniline, 99%

Catalog No. AAA1474622
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1474622 100 g
AAA1474614 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1474622 Supplier Thermo Scientific Chemicals Supplier No. A1474622
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 538-51-2 | C13H11N | 181.24 g/mol

N-Benzylideneaniline is an additional stain which is used in various immunoassays including flow cytometry, immunofluorescense, immunohistochemistry, and other applications.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Benzylideneaniline is an additional stain which is used in various immunoassays including flow cytometry, immunofluorescense, immunohistochemistry, and other applications.

Solubility
Soluble in water (partly), methanol, chloroform, alcohol, and acetic anhydride.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents.

Chemical Identifiers

CAS 538-51-2
Molecular Formula C13H11N
Molecular Weight (g/mol) 181.24
MDL Number MFCD00003027
InChI Key UVEWQKMPXAHFST-UHFFFAOYSA-N
Synonym n-benzylideneaniline, benzalaniline, benzylideneaniline, e-n-benzylideneaniline, n-benzylidenaniline, n-benzalaniline, benzaldehyde anil, aniline, n-benzylidene, benzenamine, n-phenylmethylene, n-phenylmethylene benzenamine
PubChem CID 10858
IUPAC Name N,1-diphenylmethanimine
SMILES C(=NC1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 52°C to 54°C
Flash Point >110°C (230°F)
Quantity 100 g
Merck Index 14,1138
Solubility Information Soluble in water (partly),methanol,chloroform,alcohol,and acetic anhydride.
Formula Weight 181.24
Percent Purity 99%
Chemical Name or Material N-Benzylideneaniline
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 208-694-9
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only