accessibility menu, dialog, popup

UserName

CAS RN 5368-81-0

IUPAC Name: methyl 3-methoxybenzoate
Synonyms: methyl 3-methoxybenzoate
Molecular Weight (g/mol): 166.18
Molecular Formula: C9H10O3
InChi Key: DUKYPQBGYRJVAN-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC=CC(OC)=C1
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results