missing translation for 'onlineSavingsMsg'
Learn More

Methyl m-Anisate 98.0+%, TCI America™

Catalog No. A138025G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A138025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A138025G Supplier TCI America Supplier No. A138025G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 5368-81-0
Molecular Formula C9H10O3
Molecular Weight (g/mol) 166.18
MDL Number MFCD00017193
InChI Key DUKYPQBGYRJVAN-UHFFFAOYSA-N
Synonym methyl m-anisate, methyl m-methoxybenzoate, 3-methoxybenzoic acid methyl ester, m-anisic acid, methyl ester, benzoic acid, 3-methoxy-, methyl ester, 3-methoxybenoic acid methyl ester, m-methoxybenzoic acid methyl ester, mehtyl 3-methoxybenzoate, acmc-1asq5, m-anisic acid methyl ester
PubChem CID 79332
IUPAC Name methyl 3-methoxybenzoate
SMILES COC(=O)C1=CC=CC(OC)=C1

Specifications

Color Colorless
Boiling Point 238°C
Quantity 25 g
Formula Weight 166.18
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Methyl m-Anisate