accessibility menu, dialog, popup

UserName

CAS RN 5350-41-4

IUPAC Name: benzyltrimethylazanium bromide
Synonyms: benzyltrimethyl ammonium bromide
Molecular Weight (g/mol): 230.15
Molecular Formula: C10H16BrN
InChi Key: UUZYBYIOAZTMGC-UHFFFAOYSA-M
SMILES: [Br-].C[N+](C)(C)CC1=CC=CC=C1
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (1)
  • (2)
Physical Form 
  • (3)
Quantity 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (3)
  • (1)
  • (2)