missing translation for 'onlineSavingsMsg'
Learn More

Benzyltrimethylammonium Bromide 98.0+%, TCI America™

Catalog No. B298025G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B298025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B298025G Supplier TCI America Supplier No. B298025G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 5350-41-4
Molecular Formula C10H16BrN
Molecular Weight (g/mol) 230.149
MDL Number MFCD00011780
InChI Key UUZYBYIOAZTMGC-UHFFFAOYSA-M
Synonym benzyltrimethylammonium bromide, n,n,n-trimethyl-1-phenylmethanaminium bromide, trimethylbenzylammonium bromide, n-benzyl-n,n,n-trimethylammonium bromide, wv 562 german, benzenemethanaminium, n,n,n-trimethyl-, bromide, nsc 24, ammonium, benzyltrimethyl-, bromide, benzyltrimethylammonium bromide btm
PubChem CID 21449
IUPAC Name benzyl(trimethyl)azanium;bromide
SMILES C[N+](C)(C)CC1=CC=CC=C1.[Br-]

Specifications

Melting Point 229°C
Color White-Yellow
Quantity 25 g
Formula Weight 230.15
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Benzyltrimethylammonium Bromide
EINECSNumber (3)-2694
RTECSNumber BO8392000
TSCA No