accessibility menu, dialog, popup

UserName

OHOFFF

CAS RN 53473-36-2

IUPAC Name: 3-[4-(trifluoromethyl)phenyl]propanoic acid
Synonyms: 3-[4-(trifluoromethyl)phenyl]propanoic acid
Molecular Weight (g/mol): 218.18
Molecular Formula: C10H9F3O2
InChi Key: OEIUMLSCWINLBB-UHFFFAOYSA-N
SMILES: OC(=O)CCC1=CC=C(C=C1)C(F)(F)F
Molecular Weight (g/mol) 
  • (3)
  • (2)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (3)
  • (2)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)