missing translation for 'onlineSavingsMsg'
Learn More

4-(Trifluoromethyl)hydrocinnamic acid, 95%, Thermo Scientific™

Catalog No. p-3779778
Change view
Click to view available options
Quantity:
1g
5g
25g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC397380010 1g Glass bottle
AC397380050 5g Glass bottle
AC397380250 25g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC397380010 Supplier Thermo Scientific Chemicals Supplier No. 397380010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Chemical Identifiers

CAS 53473-36-2
Molecular Formula C10H9F3O2
Molecular Weight (g/mol) 218.17
MDL Number MFCD00674032
InChI Key OEIUMLSCWINLBB-UHFFFAOYSA-N
Synonym 4-trifluoromethyl hydrocinnamic acid, 3-4-trifluoromethyl phenyl propanoic acid, 3-4-trifluoromethyl phenyl propionic acid, benzenepropanoic acid, 4-trifluoromethyl, 3-4-trifluoromethylphenyl propionic acid, 4-trifluoromethyl phenylpropionic acid, p-trifluoromethyl benzylacetic acid, 3-4-trifluoromethyl-phenyl-propionic acid
PubChem CID 3588736
IUPAC Name 3-[4-(trifluoromethyl)phenyl]propanoic acid
SMILES C1=CC(=CC=C1CCC(=O)O)C(F)(F)F

Specifications

CAS Min % 94.0
CAS Max % 100.0
Infrared Spectrum Authentic
Assay Percent Range 94% min. (HPLC)
Packaging Glass bottle
Linear Formula COOH(CH2)2C6H4CF3
Quantity 1g
Formula Weight 218.17
Percent Purity 95%
Chemical Name or Material 4-(Trifluoromethyl)hydrocinnamic acid
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only