accessibility menu, dialog, popup

UserName

CAS RN 5337-17-7

NH2POHHOO

CAS RN 5337-17-7

IUPAC Name: (4-aminophenyl)phosphonic acid
Synonyms: phosphanilic acid
Molecular Weight (g/mol): 173.11
Molecular Formula: C6H8NO3P
InChi Key: OAOBMEMWHJWPNA-UHFFFAOYSA-N
SMILES: NC1=CC=C(C=C1)P(O)(O)=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results