CAS RN 53312-81-5
IUPAC Name:
5-amino-2-fluorobenzonitrile
Synonyms:
5-amino-2-fluorobenzonitrile
Molecular Weight (g/mol):
136.13
Molecular Formula:
C7H5FN2
InChi Key:
HHTRAISBAAXRKZ-UHFFFAOYSA-N
SMILES:
NC1=CC(C#N)=C(F)C=C1
1
–
1
of
1
results
1
Promotions Available
5-Amino-2-fluorobenzonitrile, 97%
5-Amino-2-fluorobenzonitrile, 97%
CAS: 53312-81-5 Molecular Formula: C7H5FN2 Molecular Weight (g/mol): 136.13 MDL Number: MFCD00277872 InChI Key: HHTRAISBAAXRKZ-UHFFFAOYSA-N Synonym: 3-cyano-4-fluoroaniline,2-fluoro-5-aminobenzonitrile,5-amino-2-fluoro-benzonitrile,benzonitrile, 5-amino-2-fluoro,4-fluoro-3-cyanoaniline,3-amino-6-fluorobenzonitrile,ksc494a6l,3-cyano-4-fluoroanilin,5-amino-2-fluorobenzenecarbonitrile,buttpark 179\04-11 PubChem CID: 2737673 IUPAC Name: 5-amino-2-fluorobenzonitrile SMILES: NC1=CC(C#N)=C(F)C=C1