accessibility menu, dialog, popup

UserName

CAS RN 5328-01-8

CH3O

CAS RN 5328-01-8

IUPAC Name: 1-ethoxynaphthalene
Synonyms: 1-ethoxy-naphthalin
Molecular Weight (g/mol): 172.23
Molecular Formula: C12H12O
InChi Key: APWZAIZNWQFZBK-UHFFFAOYSA-N
SMILES: CCOC1=C2C=CC=CC2=CC=C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results