missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Ethoxynaphthalene 95.0+%, TCI America™
Spécifications
Spécifications
| Quantity | 5 g |
| Formula Weight | 172.23 |
| Percent Purity | ≥95.0% (GC) |
| Physical Form | Liquid |
| Chemical Name or Material | 1-Ethoxynaphthalene |
Sécurité et manipulation
| TSCA | No |