accessibility menu, dialog, popup

UserName

CH3NNSHOH3CH2NNONOO

CAS RN 532-43-4

IUPAC Name: 3-[(4-amino-2-methylpyrimidin-5-yl)methyl]-5-(2-hydroxyethyl)-4-methyl-3H-1λ⁴,3-thiazol-1-ylium nitrate
Synonyms: thiamine nitrate
Molecular Weight (g/mol): 327.36
Molecular Formula: C12H17N5O4S
InChi Key: UIERGBJEBXXIGO-UHFFFAOYSA-N
SMILES: [O-][N+]([O-])=O.CC1=C(CCO)[S+]=CN1CC1=CN=C(C)N=C1N
Physical Form 
  • (3)
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)
  • (2)
Percent Purity 
  • (3)
  • (1)
Grade 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (2)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)