missing translation for 'onlineSavingsMsg'
Learn More

Thiamine Nitrate 98.0+%, TCI America™

Catalog No. T0182500G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T0182500G 500 g
T018225G 25 g
T0182100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. T0182500G Supplier TCI America Supplier No. T0182500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 532-43-4
Molecular Formula C12H17N5O4S
Molecular Weight (g/mol) 327.36
MDL Number MFCD00036330
InChI Key UIERGBJEBXXIGO-UHFFFAOYSA-N
Synonym thiamine nitrate, thiamine mononitrate, vitamin b1 nitrate, vitamin b1 mononitrate, aneurine mononitrate, aneurine nitrate, betabion mononitrate, thiamine nitrate salt, vitamin b sub 1 nitrate, thiamine mononitrate usp
PubChem CID 10762
IUPAC Name 3-[(4-amino-2-methylpyrimidin-5-yl)methyl]-5-(2-hydroxyethyl)-4-methyl-3H-1λ⁴,3-thiazol-1-ylium nitrate
SMILES [O-][N+]([O-])=O.CC1=C(CCO)[S+]=CN1CC1=CN=C(C)N=C1N

Specifications

Melting Point 190°C
Color White-Yellow
pH 6.5 to 8.0 (10g/L H2O 20°C)
Quantity 500 g
Formula Weight 327.36
Percent Purity ≥98.0% (HPLC,E)
Physical Form Crystalline Powder
Chemical Name or Material Thiamine Nitrate
RTECSNumber XI7400000
TSCA Yes