missing translation for 'onlineSavingsMsg'
Learn More

Thiamine Nitrate Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111014494
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111014494 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111014494 Supplier Merck Emd Millipore Supplier No. PHR13371G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Certified pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards. Thiamine Nitrate is a mononitrate salt of thiamine. It is synthesized from thiamine hydrochloride following a sequence of steps. Thiamine Nitrate is used as a dietary supplement in crackers, eggs substitute and enriched flour.

Specifications

CAS 532-43-4
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP (PhEur) and BP primary standards, where they are available.
Physical Form Neat
Linear Formula C12H17N5O4 S
MDL Number MFCD00036330
Synonym Thiamine nitrate; Vitamin B1 nitrate
Type Certified Reference Material
Quantity 1 g
Formula Weight 327.36
Grade Certified Reference Material