accessibility menu, dialog, popup

UserName

CAS RN 53164-05-9

CH3ONH3COOOHOOCl

CAS RN 53164-05-9

IUPAC Name: 2-({2-[1-(4-chlorobenzoyl)-5-methoxy-2-methyl-1H-indol-3-yl]acetyl}oxy)acetic acid
Synonyms: acemetacin acemix 阿西美辛 heumann brand of acemetacin intermuti brand of acemetacin rantudil stada brand of acemetacin mibe brand of acemetacin acemetacin - chemical docpharm brand of acemetacin
Molecular Weight (g/mol): 415.83
Molecular Formula: C21H18ClNO6
InChi Key: FSQKKOOTNAMONP-UHFFFAOYSA-N
SMILES: COC1=CC=C2N(C(=O)C3=CC=C(Cl)C=C3)C(C)=C(CC(=O)OCC(O)=O)C2=C1
Physical Form 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)